Sand, quartz 50-70 mesh, Fluka™

Supplier: Honeywell Chemicals
Danger

274739-1KG 274739-5KG 274739-500G
HONC274739-1KGEA 190 SGD
HONC274739-1KG HONC274739-5KG HONC274739-500G
Sand, quartz 50-70 mesh, Fluka™
Sand, quartz
Formula: SiO₂
MW: 60.08 g/mol
Boiling Pt: 2.210 °C (1.013 hPa)
Melting Pt: 1.710 °C
Density: 2.650 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 14808-60-7
EINECS: 238-878-4

Order Now

Specification Test Results

Particle Size 50-70 mesh
Appearance complying
Granulation 0.21-0.3 mm Min. 85 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR