Molecular sieve dehydrate, beads for drying gases, with indicator, Ø 2-5 mm, Fluka™
Supplier: Honeywell Chemicals
28211-1KG
HONC28211-1KGEA 1960 SGD
HONC28211-1KG
Molecular sieve dehydrate, beads for drying gases, with indicator, Ø 2-5 mm, Fluka™
Molecular sieve dehydrate
Order Now
Specification Test Results
Appearance (color) complying
Loss on ignition (900°C) Max. 4.0 %
Water adsorption capacity Min. 15 %
SPECIFICATIONS
Environmentally Preferable