Hexane (mixture of isomers) ≥98.5%, CHROMASOLV™ for HPLC, Riedel-de Haën™

Supplier: Honeywell Chemicals
Danger

Synonyms: Hexanes, Isohexane

293253-1L
HONC293253-1LEA 197 SGD
HONC293253-1L
Hexane (mixture of isomers) ≥98.5%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Hexane (mixture of isomers)

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency

Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 48…65 °C (1.013 hPa)
Melting Pt: –154 °C
Density: 0.66 g/cm³ (20 °C)
Flash Pt: –26 °C
Storage Temperature: Ambient
MDL Number: MFCD02179311
CAS Number: 110-54-3
UN: 1208
ADR: 3 II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c38497992e162315d0f3563bfcd3dc478422095&vid=7880d919bf16a88cca44241139a2be8054e77804&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Autoignition Temperature 437 °F
Identity (IR) complying
Assay of hexane (all isomers, GC) Min. 98.50 %
Refractive index (n 20/D) 1.377 - 1.382
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.02 %
Titrable Acid Max. 0.0003 meq/g
S-compounds (as S) Max. 0.005 %
Thiophene complying
Absorbance at 400 nm Max. 0.01
Absorbance at 250 nm Max. 0.01
Absorbance at 240 nm Max. 0.02
Absorbance at 230 nm Max. 0.05
Absorbance at 220 nm Max. 0.10
Absorbance at 210 nm Max. 0.30
Absorbance at 195 nm Max. 1.0
APHA Max. 10
Vapor Density 3 (vs air)
Explosion Limit 7.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR