Methylthymol blue sodium salt indicator for metal titration, Fluka™

Supplier: Honeywell Chemicals

Synonyms: Sodium 2,2',2'',2'''-((((1,1-dioxido-3H-benzo[c][1,2]oxathiole-3,3-diyl)bis(6-hydroxy-5-isopropyl-2-methyl-3,1-phenylene))bis(methylene))bis(azanetriyl))tetraacetate, Methylthymol Blue tetrasodium salt

31042-5G
HONC31042-5GEA 921 SGD
HONC31042-5G
Methylthymol blue sodium salt indicator for metal titration, Fluka™
Methylthymol Blue sodium salt
Formula: C₃₇H₄₀N₂Na₄O₁₃S
Storage Temperature: Ambient
MDL Number: MFCD00148905
CAS Number: 1945-77-3
EINECS: 217-743-3

Order Now

Specification Test Results

Identity (IR) complying
Appearance of the solution complying
Suitability for Complexometry complying
Absorption maximum 433 - 443 nm
Absorbtion (1%/1cm) 90 - 130


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR