Sodium chloride concentrate for dilution to 1L, 0.1 M NaCl (0.1N) volumetric solution, Fluka™

Supplier: Honeywell Chemicals

Synonyms: Saline, NaCl, Sea salt, Halite

32038-1EA
HONC32038-1EAEA 113 SGD
HONC32038-1EA
Sodium chloride concentrate for dilution to 1L, 0.1 M NaCl (0.1N) volumetric solution, Fluka™
Sodium chloride

Concentrate for 1L standard solution

Formula: NaCl
Density: 1,00 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-791-2

Order Now

Specification Test Results

Factor 0.998 - 1.002


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR