n-Hexane 99 GC_ASSAY_METHOD, Puriss. p.a. Reag. Ph. Eur., ACS analytical reagent, Riedel-de Haën™

Supplier: Honeywell Chemicals
Danger

Synonyms: Hexyl hydride, Hexanen, Hexane

32293-1L 32293-2.5L 32293-25L 32293-4X2.5L 32293-6X1L
HONC32293-1LEA 487 SGD
HONC32293-1L HONC32293-2.5L HONC32293-25L HONC32293-4X2.5L HONC32293-6X1L
n-Hexane 99 GC_ASSAY_METHOD, Puriss. p.a. Reag. Ph. Eur., ACS analytical reagent, Riedel-de Haën™
n-Hexane

ACS GENERAL USE SOLVENTS


  • Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • Suitable for less demanding applications such as routine isocratic HPLC
  • Variations and decrease issues traceable to solvent contamination
  • All solvents are tested to meet ACS general use specifications


Beilstein Number: 1730733
RTECS Number: MN9275000
UNSPSC Code: 12191502

Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 66…69 °C(1.013 hPa)
Melting Pt: –95 °C
Density: 0.660…0.668 g/cm³ (20 °C)
Flash Pt: 7.4 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Refractive Index n20/D 1.375 (lit.)
Assay (GC) Min. 99 %
Boiling range 68 - 69 °C
Boiling range 67-69 °C Min. 95 %
Density (D 20/4) 0.658 - 0.662
Density (D 20/20) 0.659 - 0.663
Refractive index (n 20/D) 1.3745 - 1.3755
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.01 %
Free acid (as CH₃COOH) Max. 0.002 %
Aluminium (Al) Max. 0.00005 %
Boron (B) Max. 0.000002 %
Barium (Ba) Max. 0.00001 %
Calcium (Ca) Max. 0.00005 %
Cadmium (Cd) Max. 0.000005 %
Cobalt (Co) Max. 0.000002 %
Chromium (Cr) Max. 0.000002 %
Copper (Cu) Max. 0.000002 %
Iron (Fe) Max. 0.00001 %
Magnesium (Mg) Max. 0.00001 %
Manganese (Mn) Max. 0.000002 %
Nickel (Ni) Max. 0.000002 %
Lead (Pb) Max. 0.00001 %
Tin (Sn) Max. 0.00001 %
Zinc (Zn) Max. 0.00001 %
Aromatic substances (as C₆H₆) Max. 0.01 %
S-compounds (as S) Max. 0.005 %
Thiophene Max. 0.0001 %
Reaction against H₂SO₄ complying
APHA Max. 10

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR