Bromophenol blue sodium salt indicator, Fluka™

Supplier: Honeywell Chemicals

Synonyms: Bromophenol blue sodium, Sodium α-(3,5-dibromo-4-oxo-2,5-cyclohexadienylidene)-α-(3,5-dibromo-4-hydroxyphenyl)toluenesulphonate, Sodium 2,6-dibromo-4-(3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)phenolate, Bromophenol blue water soluble, 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2,6-dibromophenol] S,S-dioxide sodium salt

32768-50G
HONC32768-50GEA 988 SGD
HONC32768-50G
Bromophenol blue sodium salt indicator, Fluka™
Bromophenol blue sodium salt
Formula: C₁₉H₉Br₄NaO₅S
Storage Temperature: Ambient
MDL Number: MFCD00013793
CAS Number: 34725-61-6
EINECS: 252-170-2

Order Now

Specification Test Results

Appearance complying
Identity (IR) complying
Clearness of the solution complying
Change range (pH 3-4,6) complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR