Pyridine ≥99.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™

Supplier: Honeywell Chemicals
Danger

Synonyms: Azabenzene, Azine

33501-200KG
HONC33501-200KGEA 44960 SGD
HONC33501-200KG
Pyridine ≥99.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Pyridine
Formula: C₅H₅N
MW: 79.1 g/mol
Boiling Pt: 115 °C (1.013 hPa)
Melting Pt: –42 °C
Density: 0.98 g/cm3 (20 °C)
Flash Pt: 17
Storage Temperature: Ambient
MDL Number: MFCD00011732
CAS Number: 110-86-1
UN: 1282
ADR: 3 II
Merck Index: 13,08060
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f9c5934940218027dae0d80a1f98b6d5179c0a8&vid=9f2ef46a26e488d366e82869b3acb96652b5dab1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) Min. 99.5 %
Non-volatile matter Max. 0.002 %
Water (Karl Fischer) Max. 0.1 %
Ammonia (NH₃) Max. 0.002 %
Copper (Cu) Max. 0.0005 %
Chloride (Cl) Max. 0.001 %
Sulfate (SO₄) Max. 0.001 %
KMnO₄ reducing matter (as O) complying
Solubility in water complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR