Barium chloride 0.1 M BaCl2 (0.2N) volumetric solution, Fluka™

Supplier: Honeywell Chemicals
Warning

34252-1L
HONC34252-1LEA 338 SGD
HONC34252-1L
Barium chloride 0.1 M BaCl2 (0.2N) volumetric solution, Fluka™
Barium chloride
Formula: BaCl₂
Density: 1,00 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003445
CAS Number: 10361-37-2
EINECS: 233-788-1
ADR: 6.1,III

Order Now

Specification Test Results

Factor 0.998 - 1.002


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR