Cerium(IV) sulphate 0,05 M volumetric solution, Fluka™

Supplier: Honeywell Chemicals
Danger

Synonyms: Cerium disulphate, Ceric sulphate, Cerium(IV) sulphate

34253-1L
HONC34253-1LEA 410 SGD
HONC34253-1L
Cerium(IV) sulphate 0,05 M volumetric solution, Fluka™
Cerium (IV) sulphate
Formula: Ce(SO₄)₂
MW: 332.24 g/mol
Melting Pt: 195
Storage Temperature: Ambient
MDL Number: MFCD00148852
CAS Number: 13590-82-4
UN: 3264
ADR: 8 III
Merck Index: 12,02036

Order Now

Specification Test Results

Factor 0.998 - 1.002


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR