Potassium chloride 0.2 M KCl ≥99.5% volumetric solution, Fluka™

Supplier: Honeywell Chemicals

Synonyms: Potassiumchloride

34262-1L
HONC34262-1LEA 267 SGD
HONC34262-1L
Potassium chloride 0.2 M KCl ≥99.5% volumetric solution, Fluka™
Potassium chloride
Formula: KCl
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
EINECS: 231-211-8

Order Now

Specification Test Results

Factor 0.998 - 1.002


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR