Potassium permanganate 0.01 M KMnO4 (0.05N) volumetric solution, Fluka™

Supplier: Honeywell Chemicals

Synonyms: Permanganic acid potassium salt

34266-1L
HONC34266-1LEA 357 SGD
HONC34266-1L
Potassium permanganate 0.01 M KMnO4 (0.05N) volumetric solution, Fluka™
Potassium permanganate
Formula: KMnO₄
Boiling Pt: 100 °C (1,013 hPa)
Melting Pt: 0 °C
Density: 1,0 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011364
CAS Number: 7722-64-7
EINECS: 231-760-3
ADR: 9,III

Order Now

Specification Test Results

Factor 0.998 - 1.002


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR