Magnesium sulphate 0.1 M volumetric solution, Fluka™

Supplier: Honeywell Chemicals

Synonyms: MgSO4, Magnesium sulphate anhydrous

34276-1L
HONC34276-1LEA 327 SGD
HONC34276-1L
Magnesium sulphate 0.1 M volumetric solution, Fluka™
Magnesium sulphate
Formula: MgSO₄
Storage Temperature: Ambient
MDL Number: MFCD00011110
CAS Number: 7487-88-9
EINECS: 231-298-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bc2ae189b0cbc1bfc599951528466503d0d9008&vid=63867221020ebd2c8c227492e609a4764639ff79&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Factor 0.998 - 1.002


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR