Sodium carbonate 0.5 M Na2CO3 (1N) Volumetric, Fluka™

Supplier: Honeywell Chemicals

Synonyms: Calcined soda, Carbonic acid disodium salt

34277-1L
HONC34277-1LEA 264 SGD
HONC34277-1L
Sodium carbonate 0.5 M Na2CO3 (1N) Volumetric, Fluka™
Sodium carbonate
Formula: Na₂CO₃
MW: 105.99 g/mol
Boiling Pt: 1600
Melting Pt: 854
Storage Temperature: Ambient
MDL Number: MFCD00003494
CAS Number: 497-19-8
EINECS: 415-456-6
Merck Index: 14,08596
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6ff30f1487cdef86add98b38bfa8eb3dac14f6&vid=c5b129b7acb5c897b9d970ea3130d61fb1934b54&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Factor 0.998 - 1.002


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR