n-Hexane ≥95% (by GC), CHROMASOLV™ for pesticide residue analysis, for analysis of dioxins, furans and PCB, Riedel-de Haën™

Supplier: Honeywell Chemicals
Danger

Synonyms: Hexyl hydride, Hexanen, Hexane

34412-2.5L
HONC34412-2.5LEA 438 SGD
HONC34412-2.5L
n-Hexane ≥95% (by GC), CHROMASOLV™ for pesticide residue analysis, for analysis of dioxins, furans and PCB, Riedel-de Haën™
n-Hexane

  • High purity chromatography solvents
  • Suitability tested for specified use from HPLC up to UHPLC-MS applications.

Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 66…69 °C(1.013 hPa)
Melting Pt: –95 °C
Density: 0.660…0.668 g/cm³ (20 °C)
Flash Pt: 7.4 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) Min. 95 %
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.01 %
Suitability for GC-MS of dioxines, furanes, PCPs complying
Vapor Density ~3 (vs air)
Vapor Pressure 5.2 psi ( 37.7 °C)
Refractive Index n20/D 1.375 (lit.)
Explosion Limit 7.7 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR