Hexane (mixture of isomers), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™

Supplier: Honeywell Chemicals
Danger

34493-2.5L
HONC34493-2.5LEA 537 SGD
HONC34493-2.5L
Hexane (mixture of isomers), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Hexane (mixture of isomers)

  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent

Formula: H₃C(CH₂)₄CH₃
Boiling Pt: 48…65 °C (1,013 hPa)
Melting Pt: –154 °C
Density: 0,66 g/cm³ (20 °C)
Flash Pt: –26 °C
Storage Temperature: Ambient
MDL Number: MFCD02179311
CAS Number: 110-54-3
EINECS: 295-570-2
UN: 1208
ADR: 3,II
REACH: 01-2119484651-34
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c38497992e162315d0f3563bfcd3dc478422095&vid=7880d919bf16a88cca44241139a2be8054e77804&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay of isomers (GC) Min. 85 %
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.01 %
Suitability for residue analysis Max. 5 ng/l
n-Hexane (GC) Max. 4.9 %
Explosion Limit 7.5 %
Autoignition Temperature 437 °F


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR