Phenolphthalein 2% in ethanol indicator, Fluka™

Supplier: Honeywell Chemicals
Danger

Synonyms: 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one

34605-1L
HONC34605-1LEA 1430 SGD
HONC34605-1L
Phenolphthalein 2% in ethanol indicator, Fluka™
Phenolphthalein
Formula: C₂₀H₁₄O₄
Boiling Pt: 80 °C (1,013 hPa)
Density: 0,900 g/cm3 (20 °C)
Flash Pt: 24 °C
Storage Temperature: Ambient
MDL Number: MFCD00005913
CAS Number: 77-09-8
EINECS: 201-004-7
UN: 1170
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d81b539a32ba7c40a473e6afbb8e0dae525d637&vid=91bf141a7ea82baf5df563f73967884d2a4b5d79&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Test of sensitivity complying
Change range (pH 7.2-10.0) complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR