n-Hexane 97, CHROMASOLV™ LC-MS, Riedel-de Haën™

Supplier: Honeywell Chemicals
Danger

Synonyms: Hexyl hydride, Hexanen, Hexane

34986-1L 34986-2.5L 34986-4X2.5L 34986-6X1L
HONC34986-1LEA 400 SGD
HONC34986-1L HONC34986-2.5L HONC34986-4X2.5L HONC34986-6X1L
n-Hexane 97, CHROMASOLV™ LC-MS, Riedel-de Haën™
n-Hexane

  • For LC-MS and LC-UV trace metals analysis
  • HPLC / UV-VIS quality control
  • Low gradient baseline drift
  • Low levels of trace metals
  • No non-volatile impurities

Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 66…69 °C(1.013 hPa)
Melting Pt: –95 °C
Density: 0.660…0.668 g/cm³ (20 °C)
Flash Pt: 7.4 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Vapor Density ~3 (vs air)
Assay (GC) Min. 97 %
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.01 %
Free acid (as CH₃COOH) Max. 0.001 %
Free alkali (as NH₃) Max. 0.0005 %
Calcium (Ca) Max. 0.00001 %
Potassium (K) Max. 0.00001 %
Magnesium (Mg) Max. 0.00001 %
Sodium (Na) Max. 0.00001 %
Transmittance at 200 nm Min. 20 %
Transmittance at 225 nm Min. 80 %
Transmittance from 250 nm Min. 98 %
Suitability for HPLC-MS complying
Refractive Index n20/D 1.375 (lit.)
Vapor Pressure ~132 mmHg ( 20 °C)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR