Sodium nitrite 0.2 M NaNO2 (0.2N) volumetric solution, Fluka™

Supplier: Honeywell Chemicals

35272-1L
HONC35272-1LEA 152 SGD
HONC35272-1L
Sodium nitrite 0.2 M NaNO2 (0.2N) volumetric solution, Fluka™
Sodium nitrite
Formula: NaNO₂
Density: 1,01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011118
CAS Number: 7632-00-0

Order Now

Specification Test Results

Factor 0.998 - 1.002

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR