Oxalic acid 0.05 M (COOH)2 (0.1N) volumetric solution, Fluka™

Supplier: Honeywell Chemicals

Synonyms: Ethanedioic acid

35294-1L
HONC35294-1LEA 231 SGD
HONC35294-1L
Oxalic acid 0.05 M (COOH)2 (0.1N) volumetric solution, Fluka™
Oxalic acid
Formula: C₂H₂O₄
Boiling Pt: 100 °C
Density: 1 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00002573
CAS Number: 144-62-7
EINECS: 231-791-2
ADR: 8,III

Order Now

Specification Test Results

Factor 0.999 - 1.001

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR