Barium chloride 20% in aqueous solution, Fluka™

Supplier: Honeywell Chemicals
Warning

35323-5L
HONC35323-5LEA 483 SGD
HONC35323-5L
Barium chloride 20% in aqueous solution, Fluka™
Barium chloride
Formula: BaCl₂
Boiling Pt: ? 100 °C (1013 hPa)
Storage Temperature: Ambient
MDL Number: MFCD00003445
CAS Number: 10361-37-2
EINECS: 233-788-1
ADR: 6.1,III

Order Now

Specification Test Results

Assay 19 - 21 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR