2,6-Dichlorophenolindophenol sodium salt ≥97.0% (by argentometric analysis, on dry substance), Fluka™

Supplier: Honeywell Chemicals
Warning

Synonyms: Sodium 4-(3,5-dichloro-4-oxocyclohexa-2,5-dienylideneamino)phenoxide, Tillmans' reagent, 2,6-Dichloroindophenol sodium, 2,6-Dichloroindophenol sodium salt, 2,6-Dichloroindophenol sodium derivative

36180-5G
HONC36180-5GEA 303 SGD
HONC36180-5G
2,6-Dichlorophenolindophenol sodium salt ≥97.0% (by argentometric analysis, on dry substance), Fluka™
2,6-Dichlorophenolindophenol sodium salt
Formula: C₁₂H₆Cl₂NNaO₂
Melting Pt: >300 °C
Storage Temperature: Ambient
CAS Number: 620-45-1
EINECS: 210-640-4
Merck Index: 13,03093

Order Now

Specification Test Results

assay (HPLC) Min. 99 %
measurement uncertainty effective %
identity (NMR) complying
melting range 124 - 127 °C
Identity (IR) complying
water (Karl Fischer) Max. 5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR