Sodium hydroxide ≥97%, beads 20-40 mesh, Reagent Grade

Supplier: Honeywell Chemicals
Danger

Synonyms: Caustic soda

367176-2.5KG
HONC367176-2.5KGEA 463 SGD
HONC367176-2.5KG
Sodium hydroxide ≥97%, beads 20-40 mesh, Reagent Grade
Sodium hydroxide

  • ≥95% for general laboratory use.

Formula: NaOH
Boiling Pt: 1,390 °C (1.013 hPa)
Melting Pt: 319 °C
Density: 2,13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8,II
REACH: 01-2119457892-27
Merck Index: 14,08627

Order Now

Specification Test Results

Form beads
Assay Min. 96.5 %
Carbonate (as Na₂CO₃) Max. 2.5 %
Granulation complying
Particle Size 20-40 mesh
Vapor Pressure <18 mmHg ( 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR