Moisture test kit, Hydranal™
Supplier: Honeywell Chemicals
37858-1EA
HONC37858-1EAEA 964 SGD
HONC37858-1EA
Moisture test kit, Hydranal™
Standards Water Determination (by Karl Fischer method) Standards
Test kit for the visual water determination according to Karl Fischer without titrator.