Potassium permanganate concentrate for dilution to 1L, 1/500 M KMnO4 (0.01N) volumetric solution, Fluka™

Supplier: Honeywell Chemicals

Synonyms: Permanganic acid potassium salt

38136-1EA
HONC38136-1EAEA 192 SGD
HONC38136-1EA
Potassium permanganate concentrate for dilution to 1L, 1/500 M KMnO4 (0.01N) volumetric solution, Fluka™
Potassium permanganate

Concentrate for 1L standard solution

Formula: KMnO₄
Storage Temperature: Ambient
MDL Number: MFCD00011364
CAS Number: 7722-64-7
EINECS: 231-760-3
ADR: 9,III

Order Now

Specification Test Results

Factor 0.998 - 1.002
BAM/EMPA reference material effective
BAM/EMPA Certificate Number effective
Traceable to NIST SRM effective


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR