Magnesium sulphate concentrate for dilution to 1L, 0.1 M MgSO4 volumetric solution, Fluka™

Supplier: Honeywell Chemicals

Synonyms: MgSO4, Magnesium sulphate anhydrous

38146-1EA
HONC38146-1EAEA 238 SGD
HONC38146-1EA
Magnesium sulphate concentrate for dilution to 1L, 0.1 M MgSO4 volumetric solution, Fluka™
Magnesium sulphate
Formula: MgSO₄
Storage Temperature: Ambient
MDL Number: MFCD00011110
CAS Number: 7487-88-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bc2ae189b0cbc1bfc599951528466503d0d9008&vid=63867221020ebd2c8c227492e609a4764639ff79&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Factor 0.998 - 1.002


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR