Oxalic acid concentrate for dilution to 1L, 0.05 M (COOH)2 (0.1N) volumetric solution, Fluka™

Supplier: Honeywell Chemicals
Danger

Synonyms: Ethanedioic acid

Dose for preparation of 1 litre of solution.

38250-1EA
HONC38250-1EAEA 132 SGD
HONC38250-1EA
Oxalic acid concentrate for dilution to 1L, 0.05 M (COOH)2 (0.1N) volumetric solution, Fluka™
Oxalic acid
Formula: C₂H₂O₄
Boiling Pt: 100 °C
Density: 1,02 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00002573
CAS Number: 144-62-7
EINECS: 200-550-3
ADR: 8,III

Order Now

Specification Test Results

Factor 0.998 - 1.002


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR