n-Hexane ≥99.0%, CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™

Supplier: Honeywell Chemicals
Danger

Synonyms: Hexyl hydride, Hexanen, Hexane

52767-1L
HONC52767-1LEA 957 SGD
HONC52767-1L
n-Hexane ≥99.0%, CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
n-Hexane
Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 66…69 °C(1.013 hPa)
Melting Pt: –95 °C
Density: 0.660…0.668 g/cm³ (20 °C)
Flash Pt: 7.4 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c38497992e162315d0f3563bfcd3dc478422095&vid=7880d919bf16a88cca44241139a2be8054e77804&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Explosion Limit 7.7 %
Assay (GC) Min. 99 %
High-boiling fraction Max. 1 ppm
Refractive index (n 20/D) 1.374 - 1.376
Vapor Density ~3 (vs air)
Vapor Pressure 256 mmHg ( 37.7 °C)
Refractive Index n20/D 1.375 (lit.)
Autoignition Temperature 453 °F


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR