Water, CHROMASOLV™ for headspace gas chromatography, Riedel-de Haën™

Synonyms: Hydrogen oxide

53463-1L
HONC53463-1LEA 220 SGD
HONC53463-1L
Water, CHROMASOLV™ for headspace gas chromatography, Riedel-de Haën™
Water

Honeywell Chromasolv™ GC-headspace solvents:


  • Developed particularly for the identification and quantification of residual solvents in pharmaceuticals using GC-headspace methods described in monographs from the United States Pharmacopeia (USP) or the European Pharmacopoeia (Ph. Eur.) and following the International Conference on Harmonization of Technical Requirements for Registration of Pharmaceuticals for Human Use (ICH) guidelines
  • Very pure solvents with extremely low concentrations of the defined residual solvents

Formula: H₂O
Boiling Pt: 100 °C
Melting Pt: 0 °C
Density: 1 g/cm3
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now

Specification Test Results

Refractive Index n20/D 1.34 (lit.)
Suitability for the GC Headspace complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR