2-Methylbutane ≥99.5% (by GC), Puriss. p.a., Riedel-de Haën™

Supplier: Honeywell Chemicals
Danger

59070-1L
HONC59070-1LEA 830 SGD
HONC59070-1L
2-Methylbutane ≥99.5% (by GC), Puriss. p.a., Riedel-de Haën™
2-Methylbutane

Beilstein Number: 1730723
RTECS Number: EK4430000
UNSPSC Code: 12191502

Formula: CH₃CH₂CH(CH₃)₂
Boiling Pt: 28 °C (1,013 hPa)
Melting Pt: –160 °C
Density: 0,62 g/cm³ (20 °C)
Flash Pt: 7,6 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00009338
CAS Number: 78-78-4
EINECS: 201-142-8
UN: 1265
ADR: 3,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=498904a9f8b2618e19d2343aa74e247ec7121f00&vid=c7ed2044255d0d72af23a3fafec4ef6066d25097&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) Min. 99.5 %
Refractive index (n 20/D) 1.353 - 1.355
Water (Karl Fischer) Max. 0.05 %
Non-volatile matter Max. 0.001 %
Identity (NMR) complying
Aluminium (Al) Max. 0.5 ppm
Barium (Ba) Max. 0.1 ppm
Bismuth (Bi) Max. 0.1 ppm
Calcium (Ca) Max. 0.5 ppm
Cadmium (Cd) Max. 0.05 ppm
Cobalt (Co) Max. 0.02 ppm
Chromium (Cr) Max. 0.02 ppm
Copper (Cu) Max. 0.02 ppm
Iron (Fe) Max. 0.1 ppm
Potassium (K) Max. 0.5 ppm
Lithium (Li) Max. 0.1 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.02 ppm
Molybdenum (Mo) Max. 0.1 ppm
Sodium (Na) Max. 0.5 ppm
Nickel (Ni) Max. 0.02 ppm
Lead (Pb) Max. 0.1 ppm
Strontium (Sr) Max. 0.1 ppm
Zinc (Zn) Max. 0.1 ppm
Explosion Limit 8.3 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR