Potassium chloride 1.0 M KCl volumetric solution, Fluka™

Supplier: Honeywell Chemicals

Synonyms: Potassiumchloride

60121-1L
HONC60121-1LEA 271 SGD
HONC60121-1L
Potassium chloride 1.0 M KCl volumetric solution, Fluka™
Potassium chloride
Formula: KCl
Density: 1 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
EINECS: 231-211-8

Order Now

Specification Test Results

Factor 0.995 - 1.005


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR