Conductivity standard 111000 µS/cm (25 °C; 1 mol/L; KCl) for conductivity measurement, Fluka™
Supplier: Honeywell Chemicals
60131-500ML
HONC60131-500MLEA 2170 SGD
HONC60131-500ML
Conductivity standard 111000 µS/cm (25 °C; 1 mol/L; KCl) for conductivity measurement, Fluka™
Standards Conductivity Standards
Order Now
SPECIFICATIONS
Environmentally Preferable