Lanthanum(III) chloride hydrate ≥98.5% (by titration analysis on dried basis) (~7 mol/mol H₂O)

Supplier: Honeywell Chemicals
Danger

Synonyms: Lanthanum trichloride hydrate

61490-500G
HONC61490-500GEA 1960 SGD
HONC61490-500G
Lanthanum(III) chloride hydrate ≥98.5% (by titration analysis on dried basis) (~7 mol/mol H₂O)
Lanthanum (III) chloride hydrate
Formula: Cl₃La·H₂O
MW: 263.28
Boiling Pt: 1000
Melting Pt: 92…93 °C
Storage Temperature: Ambient
MDL Number: MFCD00149753
CAS Number: 20211-76-1
EINECS: 233-237-5
UN: 3260
ADR: 8 III

Order Now

Specification Test Results

Lanthanum chloride (LaCl₃) 64.5 - 70.0 %
Water (Karl Fischer) 32 - 36 %
Calcium (Ca) Max. 50 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 5 ppm
Potassium (K) Max. 10 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 5 ppm
Sodium (Na) Max. 10 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Zinc (Zn) Max. 5 ppm
Sulfate (SO₄) Max. 50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR