Hexane (mixture of isomers) ≥95%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™

Supplier: Honeywell Chemicals
Danger

Synonyms: Hexanes, Isohexane

650552-1L
HONC650552-1LEA 472 SGD
HONC650552-1L
Hexane (mixture of isomers) ≥95%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Hexane (mixture of isomers)

  • For HPLC, spectrophotometry, and environmental analysis
  • Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
  • Low UV impurity profile
  • Low impurity gradient LC baseline
  • Very low levels of non-volatile impurities

Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 66…69 °C(1.013 hPa)
Melting Pt: –95 °C
Density: 0.660…0.668 g/cm³ (20 °C)
Flash Pt: 7.4 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c38497992e162315d0f3563bfcd3dc478422095&vid=7880d919bf16a88cca44241139a2be8054e77804&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Vapor Pressure 256 mmHg ( 37.7 °C)
Assay (GC) Min. 95.0 %
Non-volatile matter Max. 0.0001 %
Water (Karl Fischer) Max. 0.01 %
Refractive index (n 20/D) 1.373 - 1.377
Absorbance at 400 nm Max. 0.005
Absorbance 280 nm Max. 0.005
Absorbance at 250 nm Max. 0.01
Absorbance at 220 nm Max. 0.05
Absorbance at 210 nm Max. 0.20
Absorbance at 200 nm Max. 0.50
Absorbance at 195 nm Max. 1.0
Fluorescence (chinin) at 365 nm Max. 1 ppb
IR spectrum complying
GC (as heptachlor epoxide) Max. 10 ng/l
Autoignition Temperature 453 °F
Refractive Index n20/D 1.375 (lit.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR