Sodium carbonate, anhydrous ≥99.999% (metals basis), TraceSELECT™ for trace analysis, Fluka™

Supplier: Honeywell Chemicals
Warning

71347-100G 71347-25G
HONC71347-100GEA 879 SGD
HONC71347-100G HONC71347-25G
Sodium carbonate, anhydrous ≥99.999% (metals basis), TraceSELECT™ for trace analysis, Fluka™
Sodium carbonate

TraceSELECT™ Ultra and TraceSELECT


  • Ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • Sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • The acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (µg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions

Formula: Na₂CO₃
Melting Pt: 854 °C
Density: 2,53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003494
CAS Number: 497-19-8
EINECS: 207-838-8
REACH: 01-2119485498-19
Merck Index: 14,08596
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6ff30f1487cdef86add98b38bfa8eb3dac14f6&vid=c5b129b7acb5c897b9d970ea3130d61fb1934b54&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity based on trace metals analysis Min. 99.999 %
Aluminium (Al) Max. 0.05 ppm
Arsenic (As) Max. 0.01 ppm
Barium (Ba) Max. 5 ppm
Calcium (Ca) Max. 0.1 ppm
Cadmium (Cd) Max. 0.005 ppm
Cerium (Ce) Max. 0.01 ppm
Cobalt (Co) Max. 0.005 ppm
Chromium (Cr) Max. 0.01 ppm
Copper (Cu) Max. 0.005 ppm
Europium (Eu) Max. 0.01 ppm
Iron (Fe) Max. 0.05 ppm
Mercury (Hg) Max. 5 ppb
Iridium (Ir) Max. 0.5 ppm
Potassium (K) Max. 1 ppm
Lithium (Li) Max. 0.5 ppm
Lanthanum (La) Max. 0.01 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.01 ppm
Nickel (Ni) Max. 0.01 ppm
Lead (Pb) Max. 0.01 ppm
Palladium (Pd) Max. 0.5 ppm
Platinum (Pt) Max. 0.5 ppm
Rhodium (Rh) Max. 0.5 ppm
Ruthenium (Ru) Max. 0.5 ppm
Antimony (Sb) Max. 0.01 ppm
Scandium (Sc) Max. 0.01 ppm
Selenium (Se) Max. 0.01 ppm
Samarium (Sm) Max. 0.01 ppm
Strontium (Sr) Max. 0.1 ppm
Thallium (Tl) Max. 0.01 ppm
Vanadium (V) Max. 0.1 ppm
Yttrium (Y) Max. 0.01 ppm
Ytterbium (Yb) Max. 0.01 ppm
Zinc (Zn) Max. 0.01 ppm
Chloride (Cl) Max. 10 ppm
Total N Max. 10 ppm
Phosphate (PO₄) Max. 5 ppm
Sulfate (SO₄) Max. 10 ppm
Silicate (as SiO₂) Max. 5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR