Sodium peroxide ≥95%, small beads ACS for Wurzschmitt-decomposition, Fluka™

Supplier: Honeywell Chemicals
Danger

Synonyms: di-Sodium peroxide

71883-1KG
HONC71883-1KGEA 4130 SGD
HONC71883-1KG
Sodium peroxide ≥95%, small beads ACS for Wurzschmitt-decomposition, Fluka™
Sodium peroxide
Formula: Na₂O₂
Boiling Pt: Decomposes on heating
Melting Pt: 660 °C
Density: 2,8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003497
CAS Number: 1313-60-6
EINECS: 215-209-4
UN: 1504
ADR: 5.1,I
Merck Index: 13,08728

Order Now

Specification Test Results

Assay 95.0 - 105.0 %
Silver (Ag) Max. 5 ppm
Aluminium (Al) Max. 20 ppm
Barium (Ba) Max. 5 ppm
Bismuth (Bi) Max. 5 ppm
Calcium (Ca) Max. 500 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 5 ppm
Potassium (K) Max. 200 ppm
Lithium (Li) Max. 5 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 5 ppm
Molybdenum (Mo) Max. 5 ppm
Nickel (Ni) Max. 10 ppm
Lead (Pb) Max. 5 ppm
Strontium (Sr) Max. 10 ppm
Thallium (Tl) Max. 5 ppm
Zinc (Zn) Max. 5 ppm
Heavy metals (as Pb) Max. 0.002 %
Chloride (Cl) Max. 20 ppm
Phosphate (PO₄) Max. 5 ppm
Sulfate (SO₄) Max. 10 ppm
Total N Max. 30 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR