Silica gel Rubin drying bags, beads 1.0-3.0 mm, with indicator 1/6 TE ( ~5 g/bag), Fluka™

Supplier: Honeywell Chemicals

72811-1KG
HONC72811-1KGEA 743 SGD
HONC72811-1KG
Silica gel Rubin drying bags, beads 1.0-3.0 mm, with indicator 1/6 TE ( ~5 g/bag), Fluka™
Silica gel Rubin
Formula: SiO₂
MW: 60.08 g/mol
Boiling Pt: 2.230 °C
Melting Pt: 1.600 °C
Density: 2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 112926-00-8
EINECS: 231-545-4

Order Now

Specification Test Results

Form beads
Appearance complying
Water absorbing capacity Min. 20 %
Change complying
Sieve analysis 1-3 mm Min. 95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR