Sodium chloride ≥99.5% for HPLC, Fluka™

Supplier: Honeywell Chemicals

73575-250G
HONC73575-250GEA 1530 SGD
HONC73575-250G
Sodium chloride ≥99.5% for HPLC, Fluka™
Sodium chloride
Formula: NaCl
Boiling Pt: 1,465 °C (1.013 hPa)
Melting Pt: 800 °C
Density: 2,160 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599

Order Now

Specification Test Results

Assay 99.5 - 100.5 %
pH (10 %, 20°C) 5.0 - 8.0
Absorbance at 220 nm Max. 0.05
Absorbance at 230 nm Max. 0.03
Absorbance at 240 nm Max. 0.02
Absorbance at 260 nm Max. 0.01
Absorbance 500 nm Max. 0.01
Appearance of the solution (c=10%) complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR