Cesium chloride, anhydrous ≥99.9%, Reagent Grade, free-flowing, Fluka™

Supplier: Honeywell Chemicals
Warning

Synonyms: Cesium Chloride

746487-1KG
HONC746487-1KGEA 3560 SGD
HONC746487-1KG
Cesium chloride, anhydrous ≥99.9%, Reagent Grade, free-flowing, Fluka™
Cesium chloride

  • ≥95% for general laboratory use.

Formula: CsCl
Melting Pt: 642 °C
Density: 3,98 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010955
CAS Number: 7647-17-8
EINECS: 231-600-2

Order Now

Specification Test Results

Assay 99 - 101 %
Metal trace analysis (ICP) Max. 1500 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR