Sodium sulphate, anhydrous ≥99%, Reagent Grade, free-flowing, Fluka™

Supplier: Honeywell Chemicals

Synonyms: Disodium sulphate, Bisodium sulphate, Sulphuric acid disodium salt

793531-12KG
HONC793531-12KGEA 1900 SGD
HONC793531-12KG
Sodium sulphate, anhydrous ≥99%, Reagent Grade, free-flowing, Fluka™
Sodium sulphate

  • ≥95% for general laboratory use.

Formula: Na₂SO₄
Melting Pt: 884 °C
Density: 2,68 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003504
CAS Number: 7757-82-6
EINECS: 231-820-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a4977cb8ca54638f22a60675bfbef71bbb77ab6&vid=942501aaa5afc8d85b82628854aac94d517322eb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 99.0 %
Solution 10 g to 100 mL (H₂O) complying
Loss on drying (130°C) Max. 0.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR