Benzyl alcohol 99,9 GC_ASSAY_METHOD, CHROMASOLV™ GC_HS, Riedel-de Haën™

Supplier: Honeywell Chemicals
Warning

Synonyms: Benzenemethanol, Phenylmethanol, Phenylcarbinol

80708-1L
HONC80708-1LEA 1080 SGD
HONC80708-1L
Benzyl alcohol 99,9 GC_ASSAY_METHOD, CHROMASOLV™ GC_HS, Riedel-de Haën™
Benzyl alcohol

Honeywell Chromasolv™ GC-headspace solvents:


  • Developed particularly for the identification and quantification of residual solvents in pharmaceuticals using GC-headspace methods described in monographs from the United States Pharmacopeia (USP) or the European Pharmacopoeia (Ph. Eur.) and following the International Conference on Harmonization of Technical Requirements for Registration of Pharmaceuticals for Human Use (ICH) guidelines
  • Very pure solvents with extremely low concentrations of the defined residual solvents

Formula: C₆H₅CH₂OH
MW: 108.14 g/mol
Boiling Pt: 205.3 °C (1.013 hPa)
Melting Pt: –15.4 °C
Density: 1.045 g/cm³ (20 °C)
Flash Pt: 100.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00004599
CAS Number: 100-51-6
EINECS: 202-859-9
Merck Index: 13,01126
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0c85c958ac64f8b86c5836e49f2c4354b22886d&vid=633ad635461465914e782d5a22480c9ff8f81bf7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) Min. 99.9 %
Refractive index (n 20/D) 1.538 - 1.541
Water (Karl Fischer) Max. 0.02 %
Acetophenone (GC) Max. 0.01 %
Benzaldehyde (GC) Max. 0.01 %
Suitability for the GC Headspace complying
Vapor Pressure 3.75 mmHg ( 77 °C)
Autoignition Temperature 817 °F
Vapor Density 3.7 (vs air)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR