Cesium chloride - lanthanum chloride buffer, for flame AAS, TraceSELECT™, Fluka™
Supplier: Honeywell Chemicals
95164-250ML
HONC95164-250MLEA 1080 SGD
HONC95164-250ML
Cesium chloride - lanthanum chloride buffer, for flame AAS, TraceSELECT™, Fluka™
Buffers Ionisation Buffers
TraceSELECT™, for flame atomic absorption spectrometry.
  • Packaged in high-quality containers appropriate for each product
  • The reagents are produced and bottled under clean-room conditionsions
Lanthanum chloride (LaCl3) 99,0 - 101,0 g/l
Cesium chloride (Cscl) 99,0 - 101,0 g/l
Density (D 20/4) 1,152 - 1,176
Calcium (Ca) max. 0,5 ppm
Order Now
SPECIFICATIONS
Environmentally Preferable