Orthoboric acid ≥99.5%, Reagent Grade

Supplier: Honeywell Chemicals
Danger

B0252-2.5KG
HONCB0252-2.5KGEA 469 SGD
HONCB0252-2.5KG
Orthoboric acid ≥99.5%, Reagent Grade
ortho-Boric acid

  • ≥95% for general laboratory use.

Formula: B(OH)₃
Boiling Pt: 185 °C (1013 hPa)
Melting Pt: 171 °C
Density: 1,510 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011337
CAS Number: 10043-35-3
EINECS: 233-139-2
REACH: 01-2119486683-25
Merck Index: 13,01326
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=098dd700154daaf6338ebb270902bb2bc704f27d&vid=6eec7d529090dd4c4b966ed8cd8d057153c5a47b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (alkalimetric) Min. 99.5 %
Colour of the solution complying
Clearness of the solution complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR