Copper(II) sulphate pentahydrate ≥98.0%, Reagent Grade

Supplier: Honeywell Chemicals
Warning

Synonyms: Cupric sulphate pentahydrate, Copper sulphate pentahydrate

C7631-500G
HONCC7631-500GEA 271 SGD
HONCC7631-500G
Copper(II) sulphate pentahydrate ≥98.0%, Reagent Grade
Copper (II) sulphate pentahydrate

  • ≥95% for general laboratory use.

Formula: CuSO₄·5H₂O
Boiling Pt: Dec. (on heating)
Melting Pt: 110 °C
Density: 2,28 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149681
CAS Number: 7758-99-8
EINECS: 231-847-6
ADR: 9,III
REACH: 01-2119520566-40
Merck Index: 13,02682

Order Now

Specification Test Results

Assay Min. 98.0 %
Iron (Fe) Max. 0.01 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR