Analytical standard, Propazine-d₇, reference material, Honeywell Fluka™

Supplier: Honeywell Chemicals

R1043-10MG
HONCR1043-10MGEA 816 SGD
HONCR1043-10MG
Analytical standard, Propazine-d₇, reference material, Honeywell Fluka™
Standards Organic Standards

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results

Order Now

SPECIFICATIONS

Description Propazine-d₇
Environmentally Preferable
CAS No. 1655498-05-7
Formula C₉D₇H₉ClN₅
Melting point 213…218 °C
Storage temperature Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR