Analytical standard, Detomidine hydrochloride monohydrate, reference material, Honeywell Fluka™

Supplier: Honeywell Chemicals

R1056-10MG
HONCR1056-10MGEA 368 SGD
HONCR1056-10MG
Analytical standard, Detomidine hydrochloride monohydrate, reference material, Honeywell Fluka™
Standards Organic Standards

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results

Order Now

SPECIFICATIONS

Description Detomidine hydrochloride monohydrate
Environmentally Preferable
CAS No. 90038-00-9
Formula C₁₂H₁₄N₂ · HCl · H₂O
Melting point 157…159 °C
Storage temperature Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR