Pesticide reference standard, Chlorethoxyfos, neat, Honeywell Fluka™

Supplier: Honeywell Chemicals

R1064-25MG
HONCR1064-25MGEA 133 SGD
HONCR1064-25MG
Pesticide reference standard, Chlorethoxyfos, neat, Honeywell Fluka™
Standards Organic Standards

High quality inorganic and organic reference materials produced according to ISO 9001.


  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results

Order Now

SPECIFICATIONS

Description Chlorethoxyfos
Environmentally Preferable
CAS No. 54593-83-8
Formula C₆H₁₁Cl₄O₃PS
Boiling point 80 °C
Flash point 128 °C
Storage temperature 2...8 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR