Analytical standard, Cyflufenamid, reference material, Honeywell Fluka™

Supplier: Honeywell Chemicals

R1085-25MG
HONCR1085-25MGEA 227 SGD
HONCR1085-25MG
Analytical standard, Cyflufenamid, reference material, Honeywell Fluka™
Standards Organic Standards

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results

Order Now

SPECIFICATIONS

Description Cyflufenamid
Environmentally Preferable
CAS No. 180409-60-3
Formula C₂₀H₁₇F₅N₂O₂
Storage temperature Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR