Organic analytical standards, [D-Asp3, E-Dhb7]-Microcystin-RR solution, 10 μg/ml in methanol, reference material, Honeywell Fluka™

R1095-1ML
HONCR1095-1MLEA 788 SGD
HONCR1095-1ML
Organic analytical standards, [D-Asp3, E-Dhb7]-Microcystin-RR solution, 10 μg/ml in methanol, reference material, Honeywell Fluka™
Standards Organic Standards

High quality inorganic and organic reference materials produced according to ISO 9001.


  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results

Order Now

SPECIFICATIONS

Description [D-Asp3, E-Dhb7]-Microcystin-RR solution
Environmentally Preferable
CAS No. 202120-08-9
Formula C₄₈H₇₃N₁₃O₁₂
Freezing point –98 °C
Boiling point 64...65 °C
Flash point 11 °C
Storage temperature –20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR