Organic analytical standards, Methyl 4-hydroxybenzoate-ring-13C6 solution, 50 μg/ml in acetone, reference material, Honeywell Fluka™

Supplier: Honeywell Chemicals

R1096-1ML
HONCR1096-1MLEA 733 SGD
HONCR1096-1ML
Organic analytical standards, Methyl 4-hydroxybenzoate-ring-13C6 solution, 50 μg/ml in acetone, reference material, Honeywell Fluka™
Standards Organic Standards

High quality inorganic and organic reference materials produced according to ISO 9001.


  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results

Order Now

SPECIFICATIONS

Description Methyl 4-hydroxybenzoate-ring-¹³C₆ solution
Environmentally Preferable
CAS No. 1581694-95-2
Formula ¹³C₆C₂H₈O₃
Freezing point –95 °C
Boiling point 55...57 °C
Flash point –17 °C
Storage temperature –20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR